A&D Weighing WP:PP143 Replacement Printer Paper for A and D Weighing Printer
Representative image only.
A&D Weighing Mfr # WP:PP143 – Item # 0103883
₹4,900.00INR / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionReplacement Printer Paper for A and D Weighing Printer
More About this Item
Parts and accessories for A&D Weighing printers.
A&D WP:PP143 is a pack of 5 laboratory balances dot matrix printer paper rolls that fits A&D 8121A printer & 8121B printer