These gauges have a built-in barometer sensor which allows for the pressure reading to display in gauge pressure or absolute pressure with a simple menu selection
Fully temperature compensated accuracy
Every model measures gauge or absolute pressure
Built-in barometer
Touchscreen display
Bluetooth and USB communications
IP67 rated
Communicates with Additel's Link mobile app
ISO 17025-accredited calibration with data included
Rubber boot, USB cable, silicone tube, 3 AA batteries, battery cover removal tool, and ISO 17025 Accredited Calibration Certificate.
Key Features
Fully temperature compensated accuracy
Every model measures gauge or absolute pressure
Built-in barometer
Touchscreen display
Bluetooth and USB communications
IP67 rated
Communicates with Additel's Link mobile app
ISO 17025-accredited calibration with data included
1 year warranty
More About this Item
Additel’s ADT685 digital pressure gauges are in a class of their own with an amazingly easy to use and read touchscreen display. The all-new modern menu structure and control interface bring a new and refreshing experience to gauge work. This completely new way of interfacing with our gauges makes the ADT685 gauges a real pleasure to use. With microprocessor technology, state-of-the-art silicon pressure sensors and plenty of onboard storage capacity, these attractive and fully temperature compensated gauges will provide performance, durability, and reliability. To provide support for intrinsically safe needs, this gauge is ready to provide pressure measurements and recordings in the most demanding of environments. Each Additel 685 is also IP67 rated to protect your investment in dusty and/or wet environments.
The 3.4-inch touchscreen display comes with a protective replaceable tempered glass screen and is a refreshing update to the traditional push button or blister pad interface. With an easy to navigate menu structure and fast touchscreen response, you will find these ADT685 gauges simple yet powerful to use. The easy-to-read display isn’t just fun to use, but it’s built rugged to handle the demands of a busy technician in the field or the laboratory.
The built-in barometer is calibrated and certified to an accuracy of 55 Pa. With this unique feature, ADT685 can be a solution for three common applications: gauge pressure measurement, absolute pressure measurement, and barometric or atmosphere pressure measurement.