Bausch & Lomb Measuring Scale, General Purpose Multi-Scale
Representative image only.
Bausch & Lomb Mfr # 81-34-36 – Item # 0388211
Improve inspection measurements
  • Glass measuring scales are compatible with measuring magnifiers
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MagnificationNone
  • LensRadii from 1/16" to 3/8", angles in single degrees to 90°, 0.005" and 0.1 mm scales, and line widths of 0.001", 0.002" and 0.003"
  • Mounting TypeCompatible with Hastings Measuring Magnifiers
  • Warranty1 Year
1 Year Warranty
Key Features
  • Glass measuring scales are compatible with measuring magnifiers
  • 1 year warranty
More About this Item
Use these measuring scales with the Hastings measuring magnifiers. Scales simply screw onto the bottom of each magnifier lens.
Measuring magnifiers 03882-10 and -50 are sold separately.
This general purpose scale measures radii from 1/16" to 3/8", angles in single degrees to 90º, 0.005" and 0.1 mm scales, and line widths of 0.001", 0.002" and 0.003".