BrandTech VITLAB® Acetal Rounded Tip Forceps, 18 cm (7") L

Representative image only.
BrandTech Mfr # 68299 – Item # 0644620
Facilitates movement of smaller objects
  • Outside grooves provide optimum handling and grip
  • Good resilience
  • 6 month warranty

PRODUCT OPTIONS

₹1,900.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Overall Length (in)7
  • Overall Length (cm)18
  • MaterialAcetal
  • SterileNo
  • DescriptionAcetal Rounded Tip Forceps, 18 cm (7") L
  • Warranty6 month
6 Month Warranty

Key Features

  • Outside grooves provide optimum handling and grip
  • Good resilience
  • 6 month warranty

More About this Item

Acetal (POM) rounded end forceps facilitate movement of smaller objects. Forceps feature grooves on the inside of the tip.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat