Burkle 5383-1000 Sampling spatula, polished SS, autoclavable, 188 mm; 50 mL

Representative image only.
Burkle Mfr # 5383-1000 – Item # 0604298
Completely welded spatula free of undercuts
  • Constructed of 316L stainless steel ideal for food and pharmaceutical applications
  • Suitable for steam sterilization up to 250°F (121°C) and flame sterilization
  • High-gloss polished finish for easy cleaning
  • 15 month warranty

₹86,600.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Material316L Stainless Steel
  • SterileNo
  • Volume (mL)50
  • Length (in)11.8
  • Length (cm)30
  • Warranty15 month
15 Month Warranty

Key Features

  • Constructed of 316L stainless steel ideal for food and pharmaceutical applications
  • Suitable for steam sterilization up to 250°F (121°C) and flame sterilization
  • High-gloss polished finish for easy cleaning
  • 15 month warranty

More About this Item

This spatula is constructed of high-gloss polished stainless steel and is completely welded for enhanced durability and stability. Sturdy material and construction make this spatula ideal for use in pharmaceutical, food, and cosmetic applications. The pointed tip and V-shaped design allow the spatula to pierce directly into solid substances or sacks, and sample directly from the container.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat