Casella CEL-712/K1 Dust Detective Kit with sampling pump
Representative image only.
Casella Mfr # 206107A – Item # 8655202
Scrolling graph of real-time data for quick and easy field analysis
  • Capture real-time data with fast warm-up time and update interval
  • Instantly see changes in concentration, even while monitor is still recording
  • Calibrate to any aerosolized particulate, including composite matter, for increased sensitivity
  • 2 year warranty
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Data LoggingYes
  • Data Storage (samples)15700
  • Logging Intervals2 seconds to 10 minutes
  • Battery3 AA (included)
  • Battery Life20 hours, typical
  • Warranty2 year
Two year warrantyConformite Europeenne
Key Features
  • Capture real-time data with fast warm-up time and update interval
  • Instantly see changes in concentration, even while monitor is still recording
  • Calibrate to any aerosolized particulate, including composite matter, for increased sensitivity
  • 2 year warranty
More About this Item
This portable aerosol monitor measures levels of any airborne particulate matter, including dust, smoke, fumes, pollen, and other aerosols produced by many industries. Comfortable, ergonomic housing with detachable probe make this unit ideal for general walk-through surveys. Large graphical LCD features color-coded screens for quick navigation.
Store up to 86,000 data points or 500 discrete measurement sessions in internal memory; at intervals of 1 second to 60 minutes. Each data logging session ends with average and maximum values calculated during the logging period. Included Casella Insight software lets you configure the monitor, download and analyze data, and produce reports.
Optional dust detective system (model 86552-02) is ideal for short-term environmental monitoring. The enclosure protects the monitor in hazardous areas, so you can leave it to log data on its own. The pump actively draws an air sample inside the enclosure to the probe, and the PUF filters provide PM10, respirable (4 µm) or PM2.5 size fractions.
Model 86552-02 includes an enclosure, sampling pump, and PUF filters.