CELLTREAT Scientific Products 229001B Standard-Length Serological Pipette, 1mL, Indv. Wrap, Paper/Plastic, Sterile; 500/Cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229001B – Item # 3456709
Bold, black ink indicate positive and negative graduations
  • Virgin polystyrene is gamma irradiated for sterility
  • Available bulk bagged or individually wrapped and bulk packed in bags
  • Save your controller from vapor damage—each pipette end has a material filter plug
  • RNase/DNase free and nonpyrogenic
  • Manufactured to ASTM standard E-934 for serological pipettes
PRODUCT OPTIONS
₹33,100.00INR / Case of 200
Sugggested alternative
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (mL)1
  • Graduation Interval0.01 mL
  • Increments (mL)0.01
  • Accuracy (%)2%
  • Body MaterialPolystyrene
  • Color CodeYellow
  • Filter TypeMaterial plug
  • Package Of100 individually wrapped per bag
  • DescriptionStandard-Length Serological Pipet, 1mL, Indv. Wrap, Paper/Plastic, Sterile; 500/Cs
Sterile
Key Features
  • Virgin polystyrene is gamma irradiated for sterility
  • Available bulk bagged or individually wrapped and bulk packed in bags
  • Save your controller from vapor damage—each pipette end has a material filter plug
  • RNase/DNase free and nonpyrogenic
  • Manufactured to ASTM standard E-934 for serological pipettes
More About this Item
These sterile serological pipettes feature bold black positive and negative graduations making them easy to read while flling or dispensing. Color coded stripes and packaging enable quick volume identification.