CELLTREAT Scientific Products 229235 Standard-Length Serological Pipette, 5mL, Bulk Packed, Sterile; 500/Cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229235 – Item # 3456702
Bold, black ink indicate positive and negative graduations
  • Virgin polystyrene is gamma irradiated for sterility
  • Available bulk bagged or individually wrapped and bulk packed in bags
  • Save your controller from vapor damage—each pipette end has a material filter plug
  • Manufactured to ASTM standard E-934 for serological pipettes
PRODUCT OPTIONS
₹48,000.00INR / Case of 200
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (mL)5
  • Graduation Interval0.1 mL
  • Increments (mL)0.1
  • Accuracy (%)2%
  • Body MaterialPolystyrene
  • Color CodeBlue
  • Filter TypeMaterial plug
  • Package Of25 bulk wrapped per bag
Sterile
Key Features
  • Virgin polystyrene is gamma irradiated for sterility
  • Available bulk bagged or individually wrapped and bulk packed in bags
  • Save your controller from vapor damage—each pipette end has a material filter plug
  • Manufactured to ASTM standard E-934 for serological pipettes
More About this Item
These sterile serological pipettes feature bold black positive and negative graduations making them easy to read while flling or dispensing. Color coded stripes and packaging enable quick volume identification.