CELLTREAT Scientific Products 229341 Sterile Treated Culture Flasks with Vented Cap, 75 cm²; 100/cs

Representative image only.
CELLTREAT Scientific Products Mfr # 229341 – Item # 0193106
Angled neck design offers easy access
  • Easily view cells through clear polystyrene
  • Sterilized by gamma irradiation
  • Hydrophilic surface is ideal for consistent cell attachment
  • Thicker, more durable walls for improved stacking capability
  • Printed lot numbers
  • Vented caps feature a 0.22 µm hydrophobic membrane

PRODUCT OPTIONS

₹76,600.00INR / Case of 100

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPolystyrene
  • Capacity (mL)250
  • Cap TypeVent
  • Culture Area (cm²)75
  • DescriptionSterile Treated Culture Flasks with Vented Cap, 75 cm²; 100/cs
Sterile

Includes

Sterile vented caps.

Key Features

  • Easily view cells through clear polystyrene
  • Sterilized by gamma irradiation
  • Hydrophilic surface is ideal for consistent cell attachment
  • Thicker, more durable walls for improved stacking capability
  • Printed lot numbers
  • Vented caps feature a 0.22 µm hydrophobic membrane

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat