CELLTREAT Scientific Products 229351 Sterile Treated Culture Flasks with Vented Cap, 182 cm²; 40/cs

Representative image only.
CELLTREAT Scientific Products Mfr # 229351 – Item # 0193108
Angled neck design offers easy access
  • Easily view cells through clear polystyrene
  • Sterilized by gamma irradiation
  • Features hydrophilic surface that is deals for consistent cell attachment
  • Thicker, more durable walls for improved stacking capability
  • Printed lot numbers
  • Large neck opening for easier access
  • Vented caps feature a 0.22 µm hydrophobic membrane

PRODUCT OPTIONS

₹65,600.00INR / Case of 40

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPolystyrene
  • Capacity (mL)600
  • Cap TypeVent
  • Culture Area (cm²)182
  • DescriptionSterile Treated Culture Flasks with Vented Cap, 182 cm²; 40/cs
Sterile

Includes

Sterile vented caps.

Key Features

  • Easily view cells through clear polystyrene
  • Sterilized by gamma irradiation
  • Features hydrophilic surface that is deals for consistent cell attachment
  • Thicker, more durable walls for improved stacking capability
  • Printed lot numbers
  • Large neck opening for easier access
  • Vented caps feature a 0.22 µm hydrophobic membrane

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat