CELLTREAT Scientific Products 229500 Sterile Suspension Culture Flasks with Vented Cap, 25 mL; 200/cs

Representative image only.
CELLTREAT Scientific Products Mfr # 229500 – Item # 0193111
Angled neck design offers easy access
  • Easily view cells through clear polystyrene
  • Sterilized by gamma irradiation
  • Vented caps feature a 0.22 µm hydrophobic membrane

PRODUCT OPTIONS

₹66,100.00INR / Case of 200

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPolystyrene
  • Capacity (mL)25
  • Cap TypeVent
  • DescriptionSterile Suspension Culture Flasks with Vented Cap, 25 mL; 200/cs
Sterile

Includes

Sterile vented caps.

Key Features

  • Easily view cells through clear polystyrene
  • Sterilized by gamma irradiation
  • Vented caps feature a 0.22 µm hydrophobic membrane

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat