CELLTREAT Scientific Products 230202 AC Power Adapter and Charger for Electronic Pipet Controller; 120 VAC
Representative image only.
CELLTREAT Scientific Products Mfr # 230202 – Item # 2520078
Replacement parts for your CELLTREAT® Electronic Pipette Controller
  • Adjust speed with the turn of a dial
  • Obtain up to 2500 pipettings (up to 8 hours of use) on a single charge
PRODUCT OPTIONS
₹19,300.00INR / Case
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithCELLTREAT® Electronic Pipet Controller
  • Power (VAC)120
  • DescriptionAC Power Adapter and Charger for Electronic Pipet Controller; 120 VAC
Key Features
  • Adjust speed with the turn of a dial
  • Obtain up to 2500 pipettings (up to 8 hours of use) on a single charge
More About this Item
This CellTreat pipette controller adapts to fill and dispense 1- to 100-mL sized pipettes. Control speed with the turn of a dial, and direction with low-pressure buttons. Avoid accidental over aspiration, contamination, and damage to the controller from volatile vapors with the included filter.