CELLTREAT Scientific Products Aspirating Pipette, 5mL, bulk packed, sterile, 200/cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229255 – Item # 3456762
Sterile and disposable pipettes—ideal for simple fluid
  • Transparent, virgin polystyrene construction has been gamma irradiated for sterility
  • Choose from three sizes in either bulk packed bags or individually wrapped
  • Unmarked and unplugged pipettes are perfect for basic fluid transfers
PRODUCT OPTIONS
₹17,500.00INR / Case of 200
Sugggested alternative
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (mL)5
  • Body MaterialVirgin polystyrene
  • Package Of25 bulk wrapped per bag
  • DescriptionAspirating Pipette, 5mL, bulk packed, sterile, 200/cs
Sterile
Key Features
  • Transparent, virgin polystyrene construction has been gamma irradiated for sterility
  • Choose from three sizes in either bulk packed bags or individually wrapped
  • Unmarked and unplugged pipettes are perfect for basic fluid transfers