Cole-Parmer 1687 MiniG® Homogenizer Foam Block, 6 x 15 mL Vials; 1/EA
Representative image only.
Cole-Parmer Item # 0460701
Accommodates a full range of sample vials with an adjustable clamp
  • Optimize performance of your HG-400 MiniG® homogenizer
  • Rugged foam block holds six 15 mL polycarbonate vials
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • MaterialFoam
Key Features
  • Optimize performance of your HG-400 MiniG® homogenizer
  • Rugged foam block holds six 15 mL polycarbonate vials
More About this Item
Multiple inserts are available for a variety of homogenizing applications.
Compatible with polycarbonate vials 04576-99.