Cole-Parmer 8500 Tube Adapters, 0.5-mL Inserts; 8/Pk
Representative image only.
Cole-Parmer Item # 1741473
Keep your Stuart centrifuge running smoothly with these replacement parts
  • Make the most of your centrifuge with interchangeable rotors and inserts
  • These inserts are desgined to hold 0.5 mL microcentrifuge tubes
₹7,500.00INR / Pkg of 8
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)8500
  • Max RCF x g3884
  • Tube Sizes Accepted0.5 mL
  • Number Of Tubes Accepted1
  • DescriptionTube Adapters, 0.5-mL Inserts; 8/Pk
Key Features
  • Make the most of your centrifuge with interchangeable rotors and inserts
  • These inserts are desgined to hold 0.5 mL microcentrifuge tubes
More About this Item
These replacement parts are compatible with the Stuart® 8500 microcentrifuge, model 1741-70. Order microcentrifuge separately.