Cole-Parmer Airflow Meter Vaneometer
Representative image only.
Cole-Parmer Item # 7890012
Accessories for your Cole-Parmer ductless fume hood
  • Continuously measure velosity of air flow into hood to determine when to change filters
  • Measure low air velocities from 25 to 400 FPM
  • Easy to use, even by untrained personnel
  • 1 year warranty
PRODUCT OPTIONS
₹23,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With Ductless fume hood
  • DescriptionAirflow Meter Vaneometer
  • Warranty1 Year
1 Year Warranty
Key Features
  • Continuously measure velosity of air flow into hood to determine when to change filters
  • Measure low air velocities from 25 to 400 FPM
  • Easy to use, even by untrained personnel
  • 1 year warranty
More About this Item
Choose from a wide range of accessories for enhanced performance
The airflow meter is a durable, economically priced instrument specifically designed to simplify the measurement of low air velocities. It is accurate to ±5% of full scale to 100 FPM and ±10% from 100 FPM to the top of scale. It has a spirit level to ensure accurate readings and the large scales are easy to read and visible from both sides.