Cole-Parmer Aspirating Pipette, 1mL, Bulk Packed, Sterile; 400/CS
Representative image only.
Cole-Parmer Item # 3456766
Aspirate and dispense sensitive solutions
  • Choose size that suits your allocating needs—available in 1-, 2-, and 5-mL
  • Pipettes are without graduations and filter plugs
PRODUCT OPTIONS
₹25,000.00INR / Case of 400
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (mL)1
  • Body MaterialPolystyrene
  • Package Of50 bulk wrapped per bag
  • DescriptionAspirating Pipet, 1mL, bulk packed, sterile, 400/cs
Sterile
Key Features
  • Choose size that suits your allocating needs—available in 1-, 2-, and 5-mL
  • Pipettes are without graduations and filter plugs
More About this Item
Virgin polystyrene pipettes are sterilized by gamma irradiation and packaged individually wrapped or in bulk bags.