Cole-Parmer AZ6741 Digital Controller Temperature Probe, High-Temp

Representative image only.
Cole-Parmer Item # 3258798
Choose the right temperature probe for your Electrothermal digital heating controller
  • Ideal for measurements up to 800°C
  • Stainless steel with porcelain sheath
  • 1 year warranty
  • Previous part number AZ6741

PRODUCT OPTIONS

₹56,400.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Probe TypePT100, 2-wire, Class B
  • Max Temperature (° C)800
  • Temperature Accuracy±0.3°C
  • Tip Length (in)5.5
  • Tip Diameter (in)0.28
  • DescriptionDigital Controller Temperature Probe, High-Temp
  • Warranty1 year
1 Year Warranty

Key Features

  • Ideal for measurements up to 800°C
  • Stainless steel with porcelain sheath
  • 1 year warranty
  • Previous part number AZ6741

More About this Item

These temperature probes are suitable for use with your Electrothermal digital heating controller, 32587-00 and -01. Controller sold separately.

This probe has a maximum temperature of 800°C and is made of stainless steel with a porcelain sheath. This probe has a PT100, 2-wire, Class B sensor and is 140 mm long with a 7 mm diameter.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat