Cole-Parmer Block Heater Insert, Single Wide, Aluminum,16 mm dia Holes
Representative image only.
Cole-Parmer Item # 3661049
Interchangeable heating blocks offer excellent thermal conductivity
  • Versatile—interchangeable heating blocks accommodate various tubes and plates
  • Block heaters sold separately
  • Single wide
  • Holds 12, 16 mm dia x 47 mm L tubes
  • 1 year warranty
PRODUCT OPTIONS
₹5,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Number Of Blocks1
  • Block Number Of Openings12
  • Block TypeAluminum
  • Block - Tube / Vial Size Accepted (mm)16
  • DescriptionBlock Heater Insert, Single Wide, Aluminum,16 mm dia Holes
  • Warranty1 Year
1 Year Warranty
Key Features
  • Versatile—interchangeable heating blocks accommodate various tubes and plates
  • Block heaters sold separately
  • Single wide
  • Holds 12, 16 mm dia x 47 mm L tubes
  • 1 year warranty
More About this Item
Interchangeable aluminum heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates. Depending on the combination of blocks selected, plastic spacers (36620-67) can be used to fill a half-width recess, ensuring optimal thermal performance. Most blocks come with a separate hole to accept a thermometer.
Order a flexi-block to accommodate multiple size tubes, vials, or cuvettes at once. Unskirted PCR plates can be heated in either size flexi-block. Skirted microtiter plates, deep-well plates, and PCR plates can only be incubated in the triple-width version.