Cole-Parmer Cell Strainer, Sterile, 40 µm, Blue, Bulk Packed; 50/CS
Representative image only.
Cole-Parmer Item # 0633665
Quickly strain, isolate, and detach cells
  • Nylon mesh produces consistent, reproducible results
  • Color-coded frame for easy identification of mesh porosity
  • Designed to be inserted into top of 50-mL centrifuge tube
  • Extended gripping tab makes handling easy
  • Gamma irradiated
  • 40 µm mesh pore size
PRODUCT OPTIONS
₹37,000.00INR / Case of 50
Reliable Protection!
Cole-Parmer® ThinTouch Nitrile Gloves, AQL 1.5
530.00 - ₹14,500.00INR / Box of 100
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialNylon
  • Pore Size (micron)40
  • Housing MaterialPolypropylene
  • SterileYes
  • DescriptionCell Strainer, Sterile, 40 µm, Blue, Bulk Packed; 50/CS
  • ColorBlue
Sterile
Key Features
  • Nylon mesh produces consistent, reproducible results
  • Color-coded frame for easy identification of mesh porosity
  • Designed to be inserted into top of 50-mL centrifuge tube
  • Extended gripping tab makes handling easy
  • Gamma irradiated
  • 40 µm mesh pore size
More About this Item
Cole-Parmer® Essentials cell strainers are sterile, easy-to-use devices for isolating primary cells to consistently obtain a uniform single-cell suspension from tissues. Constructed of a durable polypropylene housing with nylon mesh filter, these cell strainers provide a faster, easier alternative to gauze or paper filtration. The elongated lip on the housing enable aseptic handling with forceps. Sterile strainers are gamma irradiated to eliminate the risk of cross contamination.
Order convex head pestle with molded textured surface to better manipulate material in the strainer. Pestle sold separately.