Cole-Parmer Fitting, 316L Stainless Steel, Straight, Concentric Sanitary Clamp Reducer, 1-1/2" x 1"
Representative image only.
Cole-Parmer Item # 5011801
Great for food, beverage, and dairy applications—fittings comply with FDA and 3A standards
  • Ideal for general-purpose, food, beverage, and pharmaceutical applications
  • Free technical application support available to assist with tubing selection
  • 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases
  • Common centerline—useful when cavitation is present
  • 1-1/2" x 1" clamp sizes
  • 1 year warranty
PRODUCT OPTIONS
₹5,900.00INR / Each
Create a tight seal with sanitary gaskets.
Cole-Parmer® Sanitary Gaskets, Black FKM
4,500.00 - ₹7,200.00INR / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting Material316L Stainless Steel
  • Number Of Ports2
  • Connection TypeSanitary
  • Sanitary Clamp Size (Port 1) (in)1-1/2
  • Sanitary Clamp Size (Port 2) (in)1
  • Flow PatternStraight
  • ColorMetallic
  • Regulatory Approvals3A;FDA
  • DescriptionFitting, 316L Stainless Steel, Straight, Concentric Sanitary Clamp Reducer, 1-1/2" x 1"
  • Warranty1 Year
1 Year Warranty
Key Features
  • Ideal for general-purpose, food, beverage, and pharmaceutical applications
  • Free technical application support available to assist with tubing selection
  • 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases
  • Common centerline—useful when cavitation is present
  • 1-1/2" x 1" clamp sizes
  • 1 year warranty
More About this Item
Use these 316L stainless steel sanitary fittings in the food, beverage, and pharmaceutical industries. They provide excellent chemical compatibility and corrosion resistance while offering a leak-proof connection; use with our SS clamp for a secure seal. Easily remove to clean or use Clean-in-Place (CIP) process to reduce the chances of dirt and bacteria contaminating the process.