Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Hose Barb Adapter, 3/4" Clamp x 1/4" ID
Representative image only.
Cole-Parmer Item # 5011808
Great for food, beverage, and dairy applications—fittings comply with FDA and 3A standards
  • Ideal for general-purpose, food, beverage, and pharmaceutical applications
  • Free technical application support available to assist with tubing selection
  • 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases
  • Hose barb connection helps easily transition to tubing
  • 3/4" clamp size x 1/4" hose barb
  • 1 year warranty
PRODUCT OPTIONS
₹4,400.00INR / Each
Order clamps to connect sanitary clamp fittings
SPECS & DESCRIPTION
Specifications & Description
  • Fitting Material316L Stainless Steel
  • Connection TypeSanitary x Hose Barb
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 2) (in)1/4
  • Sanitary Clamp Size (Port 1) (in)3/4
  • Flow PatternStraight
  • ColorMetallic
  • Regulatory Approvals3A;FDA
  • DescriptionFitting, 316L Stainless Steel, Straight, Sanitary Clamp to Hose Barb Adapter, 3/4" Clamp x 1/4" ID
  • Warranty1 Year
1 Year Warranty
Key Features
  • Ideal for general-purpose, food, beverage, and pharmaceutical applications
  • Free technical application support available to assist with tubing selection
  • 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases
  • Hose barb connection helps easily transition to tubing
  • 3/4" clamp size x 1/4" hose barb
  • 1 year warranty
More About this Item
Use these 316L stainless steel sanitary fittings in the food, beverage, and pharmaceutical industries. They provide excellent chemical compatibility and corrosion resistance while offering a leak-proof connection; use with our SS clamp for a secure seal. Easily remove to clean or use Clean-in-Place (CIP) process to reduce the chances of dirt and bacteria contaminating the process.