Cole-Parmer FLASK UNDERLAY Neoprene Underlay Support for 250 or 500 mL Flask, 1/Pk
Representative image only.
Cole-Parmer Item # 3455355
₹7,700.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNeoprene
More About this Item
Flask support holds one 250- or 500-mL round or reaction bottom flask. Neoprene material withstands temperatures to 350°F (175°C). Outer diameter measures 5 1⁄2" and inner diameter measures 2 1⁄4".