Cole-Parmer Flexi-Block Insert with Beads, Triple Wide
Representative image only.
Cole-Parmer Item # 3662091
Interchangeable heating blocks offer excellent thermal conductivity
  • Versatile—interchangeable heating blocks accommodate various tubes and plates
  • Block heaters sold separately
  • Heat various size tubes, vials, or cuvettes at once
  • Triple wide—measures 95 x 225 x 50 mm
  • 44 mm deep
  • 1 year warranty
PRODUCT OPTIONS
₹31,200.00INR / Each
OFTEN BOUGHT WITH…
Cole-Parmer® BH-250 Series Dry-Block Heaters
143,000.00 - ₹241,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Number Of Blocks3
  • Block Number Of Openings1
  • Block TypeCeramic beads/Aluminum
  • Warranty1 Year
1 Year Warranty
Key Features
  • Versatile—interchangeable heating blocks accommodate various tubes and plates
  • Block heaters sold separately
  • Heat various size tubes, vials, or cuvettes at once
  • Triple wide—measures 95 x 225 x 50 mm
  • 44 mm deep
  • 1 year warranty
More About this Item
Interchangeable aluminum heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates. Depending on the combination of blocks selected, plastic spacers (36620-67) can be used to fill a half-width recess, ensuring optimal thermal performance. Most blocks come with a separate hole to accept a thermometer.
Order a flexi-block to accommodate multiple size tubes, vials, or cuvettes at once. Unskirted PCR plates can be heated in either size flexi-block. Skirted microtiter plates, deep-well plates, and PCR plates can only be incubated in the triple-width version.
Flexi-blocks use high-density autoclavable ceramic beads that exhibit high mechanical strength. Triple-width flexi-block insert accepts unskirted PCR plates as well as skirted microtiter plates, deep-well plates, and PCR plates.