Cole-Parmer Flow Through Tubing Weight, PTFE, 3/16" to 1/4" Tubing
Representative image only.
Cole-Parmer Item # 5011798
Prevent tubing from coiling or floating during fluid transfer applications
  • Tapered hose barb fitting easily inserts into the inner diameter (ID) of flexible tubing
  • Keep tubing submerged in liquid, preventing floatation, and ensuring uninterrupted fluid flow
  • Constructed of chemically inert PTFE
  • Autoclavable—allowing for easy sterilization and reuse
  • Suitable for tubing with 3/16" to 1/4" inner diameter
PRODUCT OPTIONS
₹9,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Minimum ID (in)0.1875
  • Maximum ID (in)0.25
  • Width (InchShort)9/16
  • Height (InchShort)1 25/64
  • Minimum ID (mm)4.7625
  • Maximum ID (mm)6.35
  • DescriptionFlow Through Tubing Weight, PTFE, 3/16" to 1/4" Tubing
Key Features
  • Tapered hose barb fitting easily inserts into the inner diameter (ID) of flexible tubing
  • Keep tubing submerged in liquid, preventing floatation, and ensuring uninterrupted fluid flow
  • Constructed of chemically inert PTFE
  • Autoclavable—allowing for easy sterilization and reuse
  • Suitable for tubing with 3/16" to 1/4" inner diameter
More About this Item
Cole-Parmer® flow-through tubing weights provide the perfect solution for keeping tubing submerged, preventing floatation, and ensuring uninterrupted flow throughout the fluid transfer process. Crafted from high-quality PTFE, these tubing weights are chemically inert, ensuring compatibility with a wide range of fluids and preventing contamination. Weighted ends are open in the center and feature a tapered hose barb fitting for easy attachment to flexible tubing , ensuring a snug and secure fit. Designed for maximum convenience and versatility, our tubing weights are autoclavable at 121°C for 20 minutes at 15 psi.