Cole-Parmer Glass Media Bottles, Class A, Round, 1000 mL (33.8 oz); 10/Pk

Representative image only.
Cole-Parmer Item # 3450341
Class A round glass media storage bottles ideal for mixing, sampling, and storing mixtures, reagents, solvents, and solutions
  • Wide-mouth opening for easy pouring
  • Resistant to corrosion by most chemicals
  • Withstand temperatures from -50 to 550 °C (-58 to 1022 °F)
  • Autoclavable PP cap and bottle
  • 5 year warranty

PRODUCT OPTIONS

₹27,100.00INR / Case of 10

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)1000
  • MaterialBorosilicate Glass
  • Min Graduation (mL)100
  • Max Graduation (mL)900
  • Graduation Interval (mL)100
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size45 mm
  • Cap ColorBlue
  • Bottle ColorClear
  • AutoclavableYes
  • Withstands High Temperature (° F)1022
  • Withstands Low Temperature (° F)-58
  • Neck Inside Diameter (ID) (mm)32
  • Neck Inside Diameter (ID) (in)1 1/4
  • Outside Diameter (OD) (in)3 15/16
  • Outside Diameter (OD) (cm)10
  • Overall Height (in)8 55/64
  • Overall Height (cm)22.5
  • DescriptionGlass Media Bottles, Class A, Round, 1000 mL (33.8 oz); 10/CS
  • Withstands High Temperature (DegreeC)550
  • Withstands Low Temperature (DegreeC)-50
  • Warranty5 years
5 Year Warranty

Key Features

  • Wide-mouth opening for easy pouring
  • Resistant to corrosion by most chemicals
  • Withstand temperatures from -50 to 550 °C (-58 to 1022 °F)
  • Autoclavable PP cap and bottle
  • 5 year warranty

More About this Item

Made of borosilicate 3.3 glass, these Class A, round media bottles provide outstanding corrosion resistance and meet ASTM E438 requirements. Able to withstand temperatures from -50 to 550 °C (-58 to 1022 °F), they have a wide mouth opening that allows for easy pouring. The clear, colorless walls have a marking spot for labeling and graduations showing approximate volume. Both the bottle and polypropylene cap are autoclavable.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat