Cole-Parmer Hollow PolyPropylene (PP) Ball Bath Covers, 10 mm; 2500/Pk
Representative image only.
Cole-Parmer Item # 0682100
Reduce vapor and heat loss by 75% with one layer of balls
  • 3 month warranty
PRODUCT OPTIONS
₹48,300.00INR / Pkg of 2500
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHollow PolyPropylene (PP) Ball Bath Covers, 10 mm; 2500/Pk
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Polypropylene balls form a tight cover over liquids to prevent change in temperature and reduce evaporation. Use instead of a bath lid with any of the baths listed in the section.