Cole-Parmer Narrow-Mouth Plastic Bottle, PFA, 60mL

Representative image only.
Cole-Parmer Item # 0605290
Provides safe storage of corrosive and hazardous chemicals
  • Withstands temperatures from -200℃ to 260℃ (-328° to 500°F)
  • Chemical and corrosion resistance
  • Smooth inner and outer surface for easy cleaning
  • Autoclavable
  • 6 month warranty

PRODUCT OPTIONS

₹9,600.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)60
  • Capacity (L)0.06
  • Capacity (oz)2.03
  • Capacity (Gal)0.016
  • MaterialPerfluoroalkoxy (PFA)
  • Cap MaterialPFA
  • Cap / Stopper Size18 mm, OD 24 mm
  • Cap Colornatural
  • Bottle ColorNatural
  • Withstands High Temperature (° F)500
  • AutoclavableYes
  • Withstands Low Temperature (° F)-328
  • Neck Inside Diameter (ID) (mm)14
  • Neck Inside Diameter (ID) (in)35/64
  • Outside Diameter (OD) (in)1 9/16
  • Outside Diameter (OD) (cm)4
  • Overall Height (in)3 3/8
  • Overall Height (cm)8.6
  • Overall Depth (in)3 5/32
  • Overall Depth (cm)8
  • Overall Width (in)1 9/16
  • Overall Width (cm)4
  • DescriptionNarrow-Mouth Plastic Bottle, PFA, 60mL
  • Warranty6 month
6 Month Warranty

Key Features

  • Withstands temperatures from -200℃ to 260℃ (-328° to 500°F)
  • Chemical and corrosion resistance
  • Smooth inner and outer surface for easy cleaning
  • Autoclavable
  • 6 month warranty

More About this Item

These perfluoroalkoxy (PFA) narrow-mouth offer excellent resistance to chemicals, high and low temperatures, and corrosion. Recommended for demanding fluid handling and storage applications. Safe for reagents which can’t be stored in metal containers.

The translucent bottles can also be used for solutions or chemicals commonly used for trace quantity analysis, ultra-trace quantity analysis, ICP-MS analysis, and isotopic analysis. They feature extremely low water-absorption, leakproof PFA caps, and non-stick surface for easy cleaning.

Before autoclaving, place cap onto bottle mouth, but do not tighten.

Certificate of Analysis
For a sample Certificate of Analysis, please go to link below under RESOURCES.
If a Lot specific Certificate of Analysis is required, please call 1-800-323-4340 for assistance.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat