Cole-Parmer Nichrome Inoculating Loops with Handles, Large, 5.5 mm ID, Red; 5/PK
Representative image only.
Cole-Parmer Item # 0439644
Nichrome wire inoculating loops used to select, inoculate, or transfer microbiological samples
  • Cools quickly after sterilization
  • Glides over agar surface smoothly without damage
  • Less expensive than platinum
  • Insulated handle to protect hand
PRODUCT OPTIONS
₹10,100.00INR / Pkg of 5
Often bought with...
Cole-Parmer® ThinTouch Nitrile Gloves, AQL 1.5
530.00 - ₹14,500.00INR / Box of 100
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Loop Inside Diameter (mm)5.5
  • B and S Wire Gauge26
  • Contained Volume (µL)5.45
  • Handle TypeInsulated Aluminum
  • ColorRed
  • DescriptionNichrome Inoculating Loops with Handles, Large, 5.5 mm ID, Red; 5/PK
Key Features
  • Cools quickly after sterilization
  • Glides over agar surface smoothly without damage
  • Less expensive than platinum
  • Insulated handle to protect hand
More About this Item
These inoculating loops, also known as streakers, have been designed to slide easily along agar surfaces without cutting or tearing. Made of twisted nichrome wire with an insulated, light-weight aluminum handle to protect the hand during use, they cool rapidly after sterilization and are more economical than platinum loops. Non-sterile and reusable, they are available calibrated (1µl & 10µl) or non-calibrated (small, medium or large). Handles are color coded for easy identification.