Cole-Parmer PCR-300-Q Thermal Cycler, Single 384-Well and Triple 96-Well Capacity; 100 to 230 VAC

Representative image only.
Cole-Parmer Item # 9394510
Easily create, modify and access programs, saving time
  • Flexible—your choice of one, two, or four block capacities in 96-well, 384-well, or mixed formats
  • Independently control each block for simplified temperature and space optimization
  • Software allows quick access to commonly used protocols
  • Save precious lab space with 4-block system—single 384-well and triple 96- well capacity
  • 3 year warranty

PRODUCT OPTIONS

Call For Price

Call For Price

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Max Temperature (° C)100
  • Min Temperature (° C)10
  • Temperature Accuracy50⁰C: ±0.25⁰C
  • Max Gradient (° C)29
  • Min Gradient (° C)1
  • Max Heating Rate3.4⁰C per second
  • Heated Lid Temperature35 to 115⁰C, or off
  • Heated Lid PressureAdjustable
  • Number of Blocks4
  • Block UniformityBlock at 55⁰C: ±0.3⁰C
  • Block DescriptionOne 384-well plate, three sets of 96 x 0.2 mL tubes or three 96-well plates
  • Memory1000 programs max
  • OutputUSB port
  • Auto Restart On Power FailureYes
  • Height (in)21 1/16
  • Width (in)27 9/16
  • Depth (in)13
  • Height (cm)53.5
  • Width (cm)70
  • Depth (cm)33
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Warranty3 Years
3 Year WarrantyConformite Europeenne

Key Features

  • Flexible—your choice of one, two, or four block capacities in 96-well, 384-well, or mixed formats
  • Independently control each block for simplified temperature and space optimization
  • Software allows quick access to commonly used protocols
  • Save precious lab space with 4-block system—single 384-well and triple 96- well capacity
  • 3 year warranty

More About this Item

These compact thermal cyclers deliver reproducible results every run—with little to no training needed. Features include a clear responsive touch screen, secure user-specific programming, adjustable heated lid, and active sample cooling for sharper amplification.

A Program Wizard generates a protocol specific to your sequence in seconds—a quick way to optimize new assays. Units also retain approximately 1000 reports for reviewing at a later date and run on an intuitive, HD Android™ tablet interface. Save your personal settings to your USB drive; sign in is as simple as inserting your USB drive, no password needed.

The four-block systems take up less space than two or four single-block units and cost less than purchasing multiple units! Control each block separately for added versatility and use by more than one person at a time, or run on the same program to increase throughput. Each block also has its own independent heated lid—no interruption of assays running at different settings.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat