Cole-Parmer pH Electrode Housing, threaded/in-line, w/o bulb guard, PVDF
Representative image only.
Oakton Item # 0599376
Simply insert electrode—screws into tee fitting
  • Pipe tee connection is 1⁄2" NPT(M)
  • Viton O-ring seals electrode for waterproof protection
  • Housing measures 313⁄16" L x 1" dia (9.7 x 2.5 cm)
  • 1 year warranty
PRODUCT OPTIONS
₹14,400.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsIn-line
  • ConnectorBNC
  • Warranty1 year
1 Year Warranty
Key Features
  • Pipe tee connection is 1⁄2" NPT(M)
  • Viton O-ring seals electrode for waterproof protection
  • Housing measures 313⁄16" L x 1" dia (9.7 x 2.5 cm)
  • 1 year warranty
More About this Item
Simply slip an electrode into this housing and screw it into a tee fitting. Pipe tee connection is 1/2'' NPT(M). Includes a VITON O-ring. KYNAR construction provides a superior chemical resistance. Maximum pressure: 150 psi. Guarded models increase electrode life. KYNAR model is suitable for harsh chemicals. Stainless steel (SS) guard models is ideal for food and high-temperature applications. Housings measures 3-13/16''L x 1'' dia.