Environmental Express pH electrode, Refillable, low maintenance
Representative image only.
Environmental Express Item # 0599181
Faster response and increased stability across a pH range of 0 to 14
  • Dome bulb offers better protection from breakage
  • Stability in low-ionic strength solutions
  • Offers resistance to Na+ ion interference
  • Compatible with most brands of pH meters (that accept standard BNC connections)
  • 6 month warranty
PRODUCT OPTIONS
₹26,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsRugged lab and field
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)100
  • ConnectorBNC
  • Warranty6 month warranty
6 Month Warranty
Key Features
  • Dome bulb offers better protection from breakage
  • Stability in low-ionic strength solutions
  • Offers resistance to Na+ ion interference
  • Compatible with most brands of pH meters (that accept standard BNC connections)
  • 6 month warranty
More About this Item
Glass body combination probes are able to withstand higher temperatures and chemical attack. All have 3.3 ft (1 m) long cables with BNC connectors.