Cole-Parmer Precision Stainless Steel Tweezers, Bent Tips, 5B-SA, 110 mm
Representative image only.
Cole-Parmer Item # 0738710
Ideal for precision handling applications
  • Antimagnetic and antiacid stainless steel construction
  • Rugged and corrosion-resistant design
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)4 1/4
  • Overall Length (cm)11
  • MaterialStainless Steel Antimagnetic
  • SterileNo
  • Style5B Bent Tips
  • DescriptionPrecision Stainless Steel Tweezers, Bent Tips, 5B-SA, 110 mm
Key Features
  • Antimagnetic and antiacid stainless steel construction
  • Rugged and corrosion-resistant design
More About this Item
These precision stainless steel tweezers offer good corrosion resistance to most chemicals, salts, and acids. Great for lab work such as handling samples or specimens, and observing them under a microscope. The antimagnetic nature of the material makes these tweezers useful in the electronic, watch, and jewelry industries.