Cole-Parmer Probe Holder for Precision Fluidized Sand Baths, Multiple Sizes
Representative image only.
Cole-Parmer Item # 1218499
  • Designed to allow free flow of media around the exterior of the holder
  • Nine 30 cm deep multi diameter size holes—1/8", 1/4", 4 mm, 5 mm, 6 mm, 7.5 mm
  • 1 year warranty
  • Previous part number FFB08PR2
PRODUCT OPTIONS
₹392,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithPrecision fluidized sand baths 12184-03 and 12184-94
  • Warranty1 year
1 Year Warranty
Key Features
  • Designed to allow free flow of media around the exterior of the holder
  • Nine 30 cm deep multi diameter size holes—1/8", 1/4", 4 mm, 5 mm, 6 mm, 7.5 mm
  • 1 year warranty
  • Previous part number FFB08PR2
More About this Item
Parts and accessories to meet your application needs and keep your sand bath operating dependably.