Cole-Parmer Reference filling solution for inner cell. 1.5M KCl gel saturated with AgCl; 125 ml
Representative image only.
Cole-Parmer Item # 0566168
Get the most out of water analysis equipment
  • Replace or upgrade electrodes, probes, cells, and other accessories
  • 1 year warranty
PRODUCT OPTIONS
₹2,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithCole-Parmer autoclavable fermentation pH electrodes
  • DescriptionReference filling solution for inner cell. 1.5M KCl gel saturated with AgCl; 125 ml
  • Warranty1 year
1 Year Warranty
Key Features
  • Replace or upgrade electrodes, probes, cells, and other accessories
  • 1 year warranty
More About this Item
Water analysis accessories for Cole-Parmer® meters. Many of these items are compatible with other brands of meter.