Cole-Parmer Removable Blue Glass Double-Junction pH Electrode, 100 Ohms RTD
Representative image only.
Cole-Parmer Item # 2700911
Easily remove electrodes in seconds without removing the holder
  • Ideal for in-line or submersion monitoring
  • 1 year warranty
PRODUCT OPTIONS
₹121,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Min pH (pH)1
  • Max pH (pH)13
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • ATC ConnectorMini-audio plug
  • ATC Element100 Ω Pt RTD
  • ConnectorBNC
  • Warranty1 year
1 Year Warranty
Key Features
  • Ideal for in-line or submersion monitoring
  • 1 year warranty
More About this Item
Double-junction electrodes prevent electrode contamination by solutions that might react with Ag+ or Cl- (heavy metals, organics, and sulfides). Use for measurements up to 13 pH. Withstands temperatures form 23 to 212ºF (-5 to 100ºC).
Note: 100Ω RTD ATC elements conform to DIN curve 43760. ATC connector is miniature audio plug.
Connector for the 1 kΩ RTD and the 3 kΩ Balco ATC elements is a spade lug.
Connector for 100 kΩ thermistor ATC elements is a standard 1/4" phone plug.