Cole-Parmer RotoFlex™ Tube Rotator Rotisserie, 10 to 15 mL Tubes; 2/PK
Representative image only.
Cole-Parmer Item # 0439738
Adapt to different racks without tools to accommodate a wide range of tubes
  • Quickly and easily switch rotisseries for enhanced versitility
  • Compatible with the RotoFlex and RotoFlex Plus systems
PRODUCT OPTIONS
₹3,300.00INR / Pkg of 2
Call For Price
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Number of Tubes7
  • Tube Size16 to 17 mm OD
  • DescriptionTube Rotator Rotisserie, 10 to 15 mL Tubes; 2/PK
Key Features
  • Quickly and easily switch rotisseries for enhanced versitility
  • Compatible with the RotoFlex and RotoFlex Plus systems
More About this Item
These rotisseries are compatible with the RotoFlex and RotoFlex Plus benchtop tube rotating shakers (sold separately). There are many paddle options to choose from based on number of tubes held as well as tube size that is accommodated.
The magnetic platform, model 04397-42, can only be used with the RotoFlex Plus system. This is ideal to optimize Western Blot hybridization. Simply attach your plastic bag containing membrane and antibody solution to the plate using the metal strips provided. These magnetic strips can be purchased separately as well as part number 04397-43.