Cole-Parmer SamplePrep 2250 Preloaded Vial Set, Precleaned Polycarbonate, 15 mL Short, 5 Vials/Set; 10 Sets/CS
Representative image only.
Item # 0457571
Preloaded with steel grinding balls for fast and convenient sample preparation
  • Optimize performance of your HG-400 MiniG® and Geno/Grinder® homogenizers
  • Each vial is loaded with two 7/16" (11 mm) steel grinding balls 04500-22
  • 6 mL working volume per vial
PRODUCT OPTIONS
₹101,000.00INR / Case of 10
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)15
  • Tube MaterialPolycarbonate
  • Special FeaturesIncludes grinding balls
Includes
50 vials with caps, 100 grinding balls, 5 foam vial holders, and 5 polyethylene carriers.
Key Features
  • Optimize performance of your HG-400 MiniG® and Geno/Grinder® homogenizers
  • Each vial is loaded with two 7/16" (11 mm) steel grinding balls 04500-22
  • 6 mL working volume per vial
More About this Item
The 4, 5, and 15 mL vial sets come in a foam vial holder within a sturdy polyethylene carrier that has the same footprint and height as a deep-well titer plate. Carrier are used in pairs and fits adjustable clamp assembly 04605-08. Run up to four vial sets at a time for high-throughput tissue homogenization and cell lysis. Use frosted PE vials 04576-97 with Cryo-Block 04500-25 to preserve temperature sensitive samples during grinding. Polyethylene is compatible with commonly used reagents such as TRIzol® solution.
Run up to four vial sets at a time for high-throughput tissue homogenization and cell lysis. Vials can be used with Cryo-Block 04575-83 to preserve temperature sensitive samples during grinding.
Each vial set comes in a foam vial holder within a sturdy polyethylene carrier that has the same footprint and height as a deep-well microplate. Carrier are used in pairs and fits adjustable clamp assembly 04605-08. Polyethylene is compatible with commonly used reagents such as TRIzol® solution.