Cole-Parmer STANDARD CHLORIDE 1000PPM
Representative image only.
Cole-Parmer Item # 2750309
Solutions needed for accurate measurements
  • Multiple ion solutions are available
  • 1 year warranty
PRODUCT OPTIONS
₹14,000.00INR / Each
Include Electrodes Today!
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 year
1 Year Warranty
Key Features
  • Multiple ion solutions are available
  • 1 year warranty
More About this Item
Choose the compatible solution kit to operate your Cole-Parmer ion-selective electrodes properly. Kits contain a 60 mL bottle of fill solution, a 475 mL bottle of standard, and a 475 mL bottle of ISA.
Individual bottles also listed.