Cole-Parmer Submersible pH Electrode, CPVC, 1 k Ohms RTD
Representative image only.
Cole-Parmer Item # 2710220
Flat measuring surface allows particles to sweep by minimizing abrasion and extending electrode life
  • Turbulent flow provides a scrubbing action which cleans the measurement surface
  • No moving parts and no need for external power simplifies installation
  • Flat surface allows viscous fluids to flow by in shear preventing build up of old material and increasing measurement accuracy
  • Extended electrode life means overall long-term cost savings
  • 1 year warranty
PRODUCT OPTIONS
₹111,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialCPVC
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • ATC Element1 kΩ Pt RTD
  • ConnectorBNC
  • DescriptionSubmersible pH Electrode, CPVC, 1 k Ohms RTD
  • Warranty1 year
1 Year Warranty
Key Features
  • Turbulent flow provides a scrubbing action which cleans the measurement surface
  • No moving parts and no need for external power simplifies installation
  • Flat surface allows viscous fluids to flow by in shear preventing build up of old material and increasing measurement accuracy
  • Extended electrode life means overall long-term cost savings
  • 1 year warranty
More About this Item
Rugged combination electrodes with flat measuring surfaces are ideal for industrial pH applications. In both coating and abrasive applications these cartridge type electrodes can improve measurement accuracy, reduce maintenance, prolong electrode life and virtually eliminate breakage. Electrodes have a unique and innovative design. In the center of the measuring surface is the pH-responsive flat glass surface. This surface is surrounded by the flat porous plastic reference junction. The large area of this porous junction has thousands of pores that provide excellent sample contact. Built into the electrode’s body is a sealed, gel-filled double junction reference design. This design provides an extra barrier against reference side contamination. Also, it allows the electrodes to be used in applications where sulfides, mercaptans, heavy metal ions and similar materials are present. The submersion assembly has a 1⁄2" MNPT thread to which a coupling and length of supporting pipe can be attached for submersion applications. It is also used when cables must be installed in conduit.