Cole-Parmer TELOS® 96-Well Filtration Microplate™, 20 µm PE, Single Fritted, Loose Wells
Representative image only.
Cole-Parmer Item # 0647974
Designed for routine filtration and sample preparation
  • Individual well is designed to ensure elution volumes are kept to a minimum
  • Includes 20 µm polyethylene frits
  • Use for gravity or vacuum application
  • Populate the base plate with only the required number of wells
PRODUCT OPTIONS
₹9,100.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (µL)600
  • DescriptionTELOS® 96-Well Filtration Microplate™, 20 µm PE, Single Fritted, Loose Wells
Key Features
  • Individual well is designed to ensure elution volumes are kept to a minimum
  • Includes 20 µm polyethylene frits
  • Use for gravity or vacuum application
  • Populate the base plate with only the required number of wells
More About this Item
Choose from loose well or populated 96-well plates to suit your application needs.
Loose wells are ideal for assays when the sample numbers vary. Order base plates 06474-60 (required) separately to use with the loose wells.