Cole-Parmer Tube Rotator Culture Tube Holder Disc, 26 mm dia Tubes
Representative image only.
Cole-Parmer Item # 0007721
Quickly and easily replace tube holder without tools
  • Choose the tube holder(s) that accommodates your tube size(s)
  • Offers a rolling action
  • Holds 30 x 26 mm tubes
  • 1 year warranty
  • Previous part number SB3/6
PRODUCT OPTIONS
₹29,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Number of Tubes30
  • Tube Size26 mm dia
  • DescriptionTube Rotator Culture Tube Holder Disc, 26 mm dia tubes
  • Warranty1 Year
1 Year Warranty
Key Features
  • Choose the tube holder(s) that accommodates your tube size(s)
  • Offers a rolling action
  • Holds 30 x 26 mm tubes
  • 1 year warranty
  • Previous part number SB3/6