Cole-Parmer Vial Cap, Seal, Aluminum, Blue, PTFE/Red Rubber Liner, 11 mm; 100/Pk
Representative image only.
Cole-Parmer Item # 9872018
Keep your samples safe with these vial accessories
  • Select from a variety of caps, septa and inserts that fit your application and needs.
  • This seal and liner is the closure only—vial sold separately
  • Limited Lifetime Warranty
PRODUCT OPTIONS
₹4,800.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeSeal
  • Cap Diameter (mm)11
  • Cap colorBlue
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Select from a variety of caps, septa and inserts that fit your application and needs.
  • This seal and liner is the closure only—vial sold separately
  • Limited Lifetime Warranty
More About this Item
Suitable for use with Cole-Parmer® Versi-Vials
Made with high quality aluminum. Use with Cole-Parmer Versi-Vials or any standard 11 mm chromatography vial. The open top allows for filling and retrieving samples with a syringe.