Cole-Parmer Vortamix™ Mini Vortexer, 100-240 VAC; International

Representative image only.
Cole-Parmer Item # 0439728
Compact and powerful enough for 50-mL centrifuge tubes
  • Press your tube to the top gasket and mix as long as needed
  • Agitate tubes at 3000 rpm with a 4.5 mm orbit—even 50-mL, conical-bottom, centrifuge tubes
  • Choose the gasket color you prefer—red is included; blue, green, and purple are available
  • 2 year warranty

PRODUCT OPTIONS

₹27,100.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Orbit Size (mm)4.5
  • Max Speed (rpm)3000
  • Capacity30-mm tube diameter
  • Number of Tubes1
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • DescriptionMini Vortexer, 100-240 VAC; International
  • Warranty2 Years
Conformite Europeenne

Key Features

  • Press your tube to the top gasket and mix as long as needed
  • Agitate tubes at 3000 rpm with a 4.5 mm orbit—even 50-mL, conical-bottom, centrifuge tubes
  • Choose the gasket color you prefer—red is included; blue, green, and purple are available
  • 2 year warranty

More About this Item

Compact vortex shaker mixes samples in centrifuge and test tubes, up to 30 mm in diameter, with little effort. The vortex motion is activated by placing a tube on the gasket cap and pressing down lightly.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat