COMARK 400YB Yellow Digital Pocket Thermometer with Boot; -58 to 400°F

Representative image only.
COMARK Mfr # 400YB – Item # 9002557
Ideal for use in the food industry and all other general applications
  • Typical response time is 30 seconds for 99% of new readings
  • 304 stainless steel penetration stem
  • NSF approved
  • 1 year warranty

PRODUCT OPTIONS

₹5,100.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Meter Measurement TypeDigital Indicator
  • Number Of Probe Channels1
  • Min Temperature (° F)-58
  • Max Temperature (° F)400
  • Min Temperature (° C)-50
  • Max Temperature (° C)200
  • Accuracy±2.0°F (±1.0°C)
  • No. of Probe(s) Included1
  • Probe Diameter (in)0.5
  • Battery1 1.5V Silver Oxide (included)
  • DescriptionLarge Head Waterproof Digital Pocket Thermometer; -58 to 400°F
  • Warranty1 year
1 Year WarrantyNSF international certified

Includes

Protective probe sheath with pocket clip and one 1.5 V button cell battery

Key Features

  • Typical response time is 30 seconds for 99% of new readings
  • 304 stainless steel penetration stem
  • NSF approved
  • 1 year warranty

More About this Item

These thermometers come in a wide range of styles for all your kitchen applications. All boast an antimicrobial coating that reduces cross-contamination.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat