COMARK KM28B Basic Food Service Thermocouple Thermometer (Probe Is Not Included)

Representative image only.
COMARK Mfr # KM28B – Item # 9002531
Ideal for use in food-related applications
  • NSF-approved
  • 1 year warranty

PRODUCT OPTIONS

₹28,800.00INR / Each

₹23,000.00 - ₹43,100.00INR / Each

for an additional
₹32,800.00INR / Each
for an additional
₹23,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels1
  • Types Of Probes AcceptedType K
  • Min Temperature (° F)-40
  • Max Temperature (° F)1000
  • Min Temperature (° C)-40
  • Max Temperature (° C)500
  • Accuracy±2.0°F (±1.0°C); ±3% of reading above 230°F (110°C) and below -22°F (30°C)
  • Resolution0.1°; 1° above 374°F (190°C)
  • Display TypeLCD
  • Height (in)5
  • Width (in)3 1/2
  • Depth (in)1 1/2
  • Battery Life300 hours
  • Battery1 9V (included)
  • Warranty1 year
1 Year WarrantyConformite Europeenne

Key Features

  • NSF-approved
  • 1 year warranty

More About this Item

Designed specially for the food industry as an easy-to-use basic everyday thermometer. This thermometer is water resistant with an easily cleanable membrane keypad. The unit produces a high degree of accuracy, with a rapid response time, and accepts a wide range of food related type K thermocouple probes with a miniature connection. Other features include unit of measure selectable, auto-off function, and protective boot.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat