COMARK SK25M Thermocouple Heavy-Duty Surface Probe with 90° Crank Shaft, Type K
Representative image only.
COMARK Mfr # SK25M – Item # 9002590
Designed specifically for use in the food industry
  • Most probes have a response time of under 0.5 seconds
  • Include a 7-foot coil cord ( except for the ATT29 probe which is 4-foot cord )
  • Measures surface temperatures of up to 1202°F (650°C)
  • Fitted with 2-pin sub-miniature connector
  • Surface probe with 90° crank shaft
  • 6 month warranty
PRODUCT OPTIONS
₹30,700.00INR / Each
₹34,000.00 - ₹43,100.00INR / Each
for an additional
₹34,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeK
  • Min Temperature (° F)-58
  • Max Temperature (° F)1202
  • Min Temperature (° C)-50
  • Max Temperature (° C)650
  • Response Time (Sec)0.4
  • Connector TypeSub-mini (2-pin)
  • Cable TypeCoiled
  • Warranty6 month
6 Month Warranty
Key Features
  • Most probes have a response time of under 0.5 seconds
  • Include a 7-foot coil cord ( except for the ATT29 probe which is 4-foot cord )
  • Measures surface temperatures of up to 1202°F (650°C)
  • Fitted with 2-pin sub-miniature connector
  • Surface probe with 90° crank shaft
  • 6 month warranty
More About this Item
Comark food probes are designed specifically for use in the food industry to ensure proper food preparation. Penetration probes are designed to be inserted directly into the food, typically poultry, beef or pork, to gauge the internal temperature and to ensure that the meat is cooked properly for safe eating. Surface probes are used to make sure the cooking surface is at the desired temperature before and during the cooking process.
This thermocouple heavy duty surface probe is fitted with a 45° crank shaft. The Type K probe is suitable for use with a range of Comark thermometers.