Corning 4091 Stripettor™ Plus Pipetting Controller, 230V

Representative image only.
Corning Mfr # 4091 – Item # 1594182
Ergonomic, lightweight, and designed for use with most brands of serological pipets
  • Precise control of aspiration and dispensing
  • Sutiable for all types of glass and plastic serological pipets from 1 mL to 100 mL
  • Settings for blow out or slow “gravity” dispensing
  • Autoclavable pipette holder
  • Fast aspiration and dispensing speed at highest setting: 25 mL in 2.5 seconds

Call For Price

SPECS & DESCRIPTION

Specifications & Description

  • SterileNo
  • Min Dispensing (mL)1
  • Max Dispensing (mL)100

Key Features

  • Precise control of aspiration and dispensing
  • Sutiable for all types of glass and plastic serological pipets from 1 mL to 100 mL
  • Settings for blow out or slow “gravity” dispensing
  • Autoclavable pipette holder
  • Fast aspiration and dispensing speed at highest setting: 25 mL in 2.5 seconds

More About this Item

The unit operates on a rechargeable Lithium battery that provides 4.5 hours of continuous use. A battery indicator light lets the user know when it’s time to recharge.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat