Costar Spin-X 8161 Centrifuge Tube Filters, Spinfilter Ca 0.22um 100/cs
Representative image only.
Costar Spin-X Mfr # 8161 – Item # 0193732
Membrane-containing filter unit within a centrifuge tube
  • Removing bacteria, cells, and particles from liquids
  • DNA removal from agarose or acrylamide gels
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • MembraneCellulose Acetate
  • Pore Size (µm)0.22
  • Tube Size (mL)2
  • Working Volume (µL)500
Key Features
  • Removing bacteria, cells, and particles from liquids
  • DNA removal from agarose or acrylamide gels
More About this Item
Ideal for HPLC sample preparation.