Costar Spin-X 8162 Centrifuge Tube Filters, Spinfilter Ca 0.45µm 96/cs

Representative image only.
Costar Spin-X Mfr # 8162 – Item # 0193734
Membrane-containing filter unit within a centrifuge tube
  • Removing bacteria, cells, and particles from liquids
  • DNA removal from agarose or acrylamide gels

PRODUCT OPTIONS

Call For Price

SPECS & DESCRIPTION

Specifications & Description

  • Tube Size (mL)2
  • Pore Size (µm)0.45
  • SterileYes
  • Working Volume (µL)500
  • MembraneCellulose Acetate
  • DescriptionCentrifuge Tube Filters, Spinfilter Ca 0.45µm 96/cs

Key Features

  • Removing bacteria, cells, and particles from liquids
  • DNA removal from agarose or acrylamide gels

More About this Item

Ideal for HPLC sample preparation.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat